Thermo Scientific Chemicals

L-Aspartic acid monosodium salt monohydrate, 99%

Catalog number: B22321.22
100 g, Each
Thermo Scientific Chemicals

L-Aspartic acid monosodium salt monohydrate, 99%

Catalog number: B22321.22
100 g, Each
Quantity
Catalog number: B22321.22
also known as B22321-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
323194-76-9
IUPAC Name
sodium 2-amino-3-carboxypropanoate hydrate
Molecular Formula
C4H8NNaO5
InChI Key
PPTHNBYUFXSJPS-UHFFFAOYNA-M
SMILES
O.[Na+].NC(CC(O)=O)C([O-])=O
Form
Powder or crystalline powder
Optical Rotation
-14.0 ± 1.0° (c=1 in water)
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%
Identification (FTIR)
Conforms

Description

L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.

Solubility
Soluble in cold water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text