Thermo Scientific Chemicals

Chromotrope FB, Thermo Scientific Chemicals

Catalog number: B22328.09
10 g, Each
Thermo Scientific Chemicals

Chromotrope FB, Thermo Scientific Chemicals

Catalog number: B22328.09
10 g, Each
Quantity
Catalog number: B22328.09
also known as B22328-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3567-69-9
IUPAC Name
disodium 4-{2-[(2E)-1-oxo-4-sulfonato-1,2-dihydronaphthalen-2-ylidene]hydrazin-1-yl}naphthalene-1-sulfonate
Molecular Formula
C20H12N2Na2O7S2
InChI Key
OJNIRNOPCQQNRY-JZFXEGLSSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)C1=C\C(=N/NC2=C3C=CC=CC3=C(C=C2)S([O-])(=O)=O)C(=O)C2=CC=CC=C12
Specific Absorbance (E 1%/1cm)
>400 (lambda-max in Distilled Water)
UV spectrophotometry
>400 (lambda-max in Distilled Water)
Appearance (Color)
Dark red or brown to black
Form
Crystalline powder or powder
Loss on Drying
<8.0% (105°C/contant wt.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text