Thermo Scientific Chemicals

4-Phenoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B22329.06
5 g, Each
Thermo Scientific Chemicals

4-Phenoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B22329.06
5 g, Each
Quantity
Catalog number: B22329.06
also known as B22329-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
67-36-7
IUPAC Name
4-phenoxybenzaldehyde
Molecular Formula
C13H10O2
InChI Key
QWLHJVDRPZNVBS-UHFFFAOYSA-N
SMILES
O=CC1=CC=C(OC2=CC=CC=C2)C=C1
Form
Liquid
Assay (GC)
≥97.5%
Free acid (titration)
<1.5%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.6090-1.6130 @ 20?C

Description

4-Phenoxybenzaldehyde was used in the synthesis of spirodiketopiperazine derivatives, benzoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenoxybenzaldehyde was used in the synthesis of spirodiketopiperazine derivatives, benzoxazoles.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text