Ethyl 5-bromothiophene-2-carboxylate, 99%, Thermo Scientific Chemicals
Ethyl 5-bromothiophene-2-carboxylate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 5-bromothiophene-2-carboxylate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22355.03
also known as B22355-03
Price (USD)/ Each
49.20
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
49.20
Add to cart
Ethyl 5-bromothiophene-2-carboxylate, 99%, Thermo Scientific Chemicals
Catalog numberB22355.03
Price (USD)/ Each
49.20
-
Add to cart
Chemical Identifiers
CAS5751-83-7
IUPAC Nameethyl 5-bromothiophene-2-carboxylate
Molecular FormulaC7H7BrO2S
InChI KeyPZNHMXAOMDQLLE-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CC=C(Br)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥98.5%
Refractive Index1.5605-1.5635 @ 20?C
FormLiquid
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromothiophene-2-carboxylate is used as an intermediate in organic syntheses

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Reuben D. Rieke.; Seung-Hoi Kim. 5-Bromo-2-pyridylzinc reagent; direct preparation and its coupling reactions. Tetrahedron Letters. 2011, 52, (2), 244-247.
  2. Hye-Soo Jung.; Hyun-Hee Cho.; Seung-Hoi Kim. Site-selective mono-oxidative addition of active zinc into carbon-bromine bond of dibrominated-thiophenes: preparation of thienylzinc reagents and their applications. Tetrahedron Letters.2013, 54, (8), 960-964.