Thermo Scientific Chemicals

trans-4-Aminocyclohexanol, 98+%, Thermo Scientific Chemicals

Catalog number: B22365.06
5 g, Each
Thermo Scientific Chemicals

trans-4-Aminocyclohexanol, 98+%, Thermo Scientific Chemicals

Catalog number: B22365.06
5 g, Each
Quantity
Catalog number: B22365.06
also known as B22365-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
27489-62-9
IUPAC Name
4-aminocyclohexan-1-ol
Molecular Formula
C6H13NO
InChI Key
IMLXLGZJLAOKJN-UHFFFAOYSA-N
SMILES
NC1CCC(O)CC1
Appearance (Color)
White to pale cream to pale brown
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥98.0%
Melting Point (clear melt)
108.0-115.0?C

Description

trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Aminocyclohexanol is used as raw material in organic synthesis and is an important intermediate in the synthesis of drugs such as Ambroxol hydrochloride. It can react with butyric acid-(2-chloro-ethyl ester) to get butyric acid 4-amino-cyclohexyl ester. This reaction needs catalytic agent Aspergillus niger lipase and solvent 2-methyl-butan-2-ol.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text