Thermo Scientific Chemicals

2-(Methylthio)naphthalene, 98%, Thermo Scientific Chemicals

Catalog number: B22368.06
5 g, Each
Thermo Scientific Chemicals

2-(Methylthio)naphthalene, 98%, Thermo Scientific Chemicals

Catalog number: B22368.06
5 g, Each
Quantity
Catalog number: B22368.06
also known as B22368-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7433-79-6
IUPAC Name
2-(methylsulfanyl)naphthalene
Molecular Formula
C11H10S
InChI Key
SSKUUDUKJMIOKQ-UHFFFAOYSA-N
SMILES
CSC1=CC=C2C=CC=CC2=C1
Appearance (Color)
White to cream or pale brown
Form
Powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
55.0-61.0?C

Description

2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)naphthalene is used to produce 1-chloro-2-methylsulfanylnaphthalene. This reaction will need reagent CuCl2-Al2O3, and the menstruum benzene

Solubility
Slightly soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text