N-Benzyloxycarbonyl-D-alaninol, 98%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-D-alaninol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-D-alaninol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B22372.14
also known as B22372-14
Price (USD)/ Each
577.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
577.00
Add to cart
N-Benzyloxycarbonyl-D-alaninol, 98%, Thermo Scientific Chemicals
Catalog numberB22372.14
Price (USD)/ Each
577.00
-
Add to cart
Chemical Identifiers
CAS61425-27-2
IUPAC Namebenzyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
Molecular FormulaC11H15NO3
InChI KeyAFPHMHSLDRPUSM-SECBINFHSA-N
SMILESC[C@H](CO)NC(=O)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Melting Point (clear melt)80.0-87.0?C
Optical Rotation5.7 ± 0.4? (C=2 in ethanol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only