Thermo Scientific Chemicals

Furfuryl mercaptan, 98+%, Thermo Scientific Chemicals

Catalog number: B22380.14
25 g, Each
Thermo Scientific Chemicals

Furfuryl mercaptan, 98+%, Thermo Scientific Chemicals

Catalog number: B22380.14
25 g, Each
Quantity
Catalog number: B22380.14
also known as B22380-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98-02-2
IUPAC Name
(furan-2-yl)methanethiol
Molecular Formula
C5H6OS
InChI Key
ZFFTZDQKIXPDAF-UHFFFAOYSA-N
SMILES
SCC1=CC=CO1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Refractive Index
1.5295-1.5335 @ 20?C

Description

Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text