Thermo Scientific Chemicals

2-(n-Hexyloxy)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: B22385.06
5 g, Each
Thermo Scientific Chemicals

2-(n-Hexyloxy)ethanol, 98%, Thermo Scientific Chemicals

Catalog number: B22385.06
5 g, Each
Quantity
Catalog number: B22385.06
also known as B22385-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
112-25-4
IUPAC Name
2-(hexyloxy)ethan-1-ol
Molecular Formula
C8H18O2
InChI Key
UPGSWASWQBLSKZ-UHFFFAOYSA-N
SMILES
CCCCCCOCCO
Refractive Index
1.4285-1.4335 @ 20°C
Appearance (Color)
Cear colorless
Form
Liquid
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

2-(n-Hexyloxy)ethanol is used as solvents in specialty printing inks and coalescing aids in surface coatings, coupling agent, rust remover, adhesives and surface cleaners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(n-Hexyloxy)ethanol is used as solvents in specialty printing inks and coalescing aids in surface coatings, coupling agent, rust remover, adhesives and surface cleaners.

Solubility
Soluble in alcohol and ether, water (9.46 g/L ).

Notes
Store under inert gas. Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text