Thermo Scientific Chemicals

2,6-Diaminopimelic acid, 96%, Thermo Scientific Chemicals

Catalog number: B22391.03
1 g, Each
Thermo Scientific Chemicals

2,6-Diaminopimelic acid, 96%, Thermo Scientific Chemicals

Catalog number: B22391.03
1 g, Each
Quantity
Catalog number: B22391.03
also known as B22391-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
583-93-7
IUPAC Name
(2R,6S)-2,6-diazaniumylheptanedioate
Molecular Formula
C7H14N2O4
InChI Key
GMKMEZVLHJARHF-SYDPRGILSA-N
SMILES
[NH3+][C@@H](CCC[C@@H]([NH3+])C([O-])=O)C([O-])=O
Appearance (Color)
White to cream to pale brown
Form
Powder and/or chunks
Assay (Non-aqueous acid-base Titration)
≥95.0 to ≤105.0%
Identification (FTIR)
Conforms

Description

2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.

Solubility
Soluble in water, diluted acids and alkali.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text