Thermo Scientific Chemicals

N-Boc-L-alaninol, 99%, Thermo Scientific Chemicals

Catalog number: B22399.14
25 g, Each
Thermo Scientific Chemicals

N-Boc-L-alaninol, 99%, Thermo Scientific Chemicals

Catalog number: B22399.14
25 g, Each
Quantity
Catalog number: B22399.14
also known as B22399-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
79069-13-9
IUPAC Name
tert-butyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
Molecular Formula
C8H17NO3
InChI Key
PDAFIZPRSXHMCO-LURJTMIESA-N
SMILES
C[C@@H](CO)NC(=O)OC(C)(C)C
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
54.0-61.0?C
Optical Rotation
-8.6 ± 0.3? (C=1.2 in methanol)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text