Thermo Scientific Chemicals

1,4-Oxazepan-5-one, 97%, Thermo Scientific Chemicals

Catalog number: B22406.06
5 g, Each
Thermo Scientific Chemicals

1,4-Oxazepan-5-one, 97%, Thermo Scientific Chemicals

Catalog number: B22406.06
5 g, Each
Quantity
Catalog number: B22406.06
also known as B22406-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10341-26-1
IUPAC Name
1,4-oxazepan-5-one
Molecular Formula
C5H9NO2
InChI Key
OXDIYMHNQAWSCL-UHFFFAOYSA-N
SMILES
O=C1CCOCCN1
Assay (GC)
≥96.0%
Water Content (Karl Fischer Titration)
≥1.5%
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Transition Temperature
79.0-88.0?C

Description

Used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a pharmaceutical intermediate.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text