3',5'-Dibromo-2'-hydroxyacetophenone, 99%, Thermo Scientific Chemicals
3',5'-Dibromo-2'-hydroxyacetophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3',5'-Dibromo-2'-hydroxyacetophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B22413.14
also known as B22413-14
Price (USD)/ Each
154.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
154.00
Add to cart
3',5'-Dibromo-2'-hydroxyacetophenone, 99%, Thermo Scientific Chemicals
Catalog numberB22413.14
Price (USD)/ Each
154.00
-
Add to cart
Chemical Identifiers
CAS22362-66-9
IUPAC Name1-(3,5-dibromo-2-hydroxyphenyl)ethan-1-one
Molecular FormulaC8H6Br2O2
InChI KeyRCKHUOIKVIOMGZ-UHFFFAOYSA-N
SMILESCC(=O)C1=CC(Br)=CC(Br)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Appearance (Color)Cream to yellow
Melting Point (clear melt)106.0-112.0°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only