Thermo Scientific Chemicals

2-Methyl-6-nitrobenzoxazole, 98%, Thermo Scientific Chemicals

Catalog number: B22415.06
5 g, Each
Thermo Scientific Chemicals

2-Methyl-6-nitrobenzoxazole, 98%, Thermo Scientific Chemicals

Catalog number: B22415.06
5 g, Each
Quantity
Catalog number: B22415.06
also known as B22415-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5683-43-2
IUPAC Name
2-methyl-6-nitro-1,3-benzoxazole
Molecular Formula
C8H6N2O3
InChI Key
DPVZLXWUFYMLBK-UHFFFAOYSA-N
SMILES
CC1=NC2=CC=C(C=C2O1)[N+]([O-])=O
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
153.0-159.0?C

Description

2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-6-nitrobenzoxazole is used as a flavoring agents.

Solubility
Soluble in oils. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text