Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B22429.14
also known as B22429-14
Price (USD)/ Each
182.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
182.00
Add to cart
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals
Catalog numberB22429.14
Price (USD)/ Each
182.00
-
Add to cart
Chemical Identifiers
CAS5909-24-0
IUPAC Nameethyl 4-chloro-2-(methylsulfanyl)pyrimidine-5-carboxylate
Molecular FormulaC8H9ClN2O2S
InChI KeySNNHLSHDDGJVDM-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CN=C(SC)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.5%
Solution Test1% in diethyl ether: Clear, colourless
Appearance (Color)White to cream
Formpowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only