Thermo Scientific Chemicals

S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: B22435.14
25 g, Each
Thermo Scientific Chemicals

S-Boc-2-mercapto-4,6-dimethylpyrimidine, 97%, Thermo Scientific Chemicals

Catalog number: B22435.14
25 g, Each
Quantity
Catalog number: B22435.14
also known as B22435-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41840-28-2
IUPAC Name
tert-butyl [(4,6-dimethylpyrimidin-2-yl)sulfanyl]formate
Molecular Formula
C11H16N2O2S
InChI Key
POTDIELOEHTPJN-UHFFFAOYSA-N
SMILES
CC1=CC(C)=NC(SC(=O)OC(C)(C)C)=N1
Appearance (Color)
Pale yellow to pale brown
Form
Crystalline powder
Elemental Analysis
Carbon C: 52-78-57.17% (Theoretical 54.98%)
Elemental Analysis
Hydrogen H: 6.44-6.98% (Theoretical 6.71%)
Elemental Analysis
Sulfur S: 12.81-13.88% (Theoretical 13.34%)

Description

S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S-Boc-2-mercapto-4,6-dimethylpyrimidine can react with propylamine to produce propyl-carbamic acid tert-butyl ester. It is used as a pharmaceutical intermediate.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text