Thermo Scientific Chemicals

1-Methyl-1-phenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: B22440.06
5 g, Each
Thermo Scientific Chemicals

1-Methyl-1-phenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: B22440.06
5 g, Each
Quantity
Catalog number: B22440.06
also known as B22440-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
618-40-6
IUPAC Name
1-methyl-1-phenylhydrazine
Molecular Formula
C7H10N2
InChI Key
MWOODERJGVWYJE-UHFFFAOYSA-N
SMILES
CN(N)C1=CC=CC=C1
Appearance (Color)
Clear yellow to orange to brown
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.5805-1.5855 @ 20?C

Description

1-Methyl-1-phenylhydrazine is used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-1-phenylhydrazine is used to make other chemicals.

Solubility
Slightly soluble in water.

Notes
Storage temperature is 2 - 8°C. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text