Perfluoro-tert-butanol, 99%, Thermo Scientific Chemicals
Perfluoro-tert-butanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perfluoro-tert-butanol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B22442.14
Price (USD)/ Each
952.65
Online exclusive
1058.00 
Save 105.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
952.65
Online exclusive
1058.00 
Save 105.35 (10%)
Add to cart
Perfluoro-tert-butanol, 99%, Thermo Scientific Chemicals
Catalog numberB22442.14
Price (USD)/ Each
952.65
Online exclusive
1058.00 
Save 105.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2378-02-1
IUPAC Name1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
Molecular FormulaC4HF9O
InChI KeyXZNOAVNRSFURIR-UHFFFAOYSA-N
SMILESOC(C(F)(F)F)(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index<1.3000 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only