Thermo Scientific Chemicals

2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22443.06
5 g, Each
Thermo Scientific Chemicals

2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22443.06
5 g, Each
Quantity
Catalog number: B22443.06
also known as B22443-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
381-98-6
IUPAC Name
2-(trifluoromethyl)prop-2-enoate
Molecular Formula
C4H2F3O2
InChI Key
VLSRKCIBHNJFHA-UHFFFAOYSA-M
SMILES
[O-]C(=O)C(=C)C(F)(F)F
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (GC)
≥97.5%
Melting Point (clear melt)
47.0-54.0?C

Description

2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text