2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals
2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B22443.14
also known as B22443-14
Price (USD)/ Each
696.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
696.00
Add to cart
2-(Trifluoromethyl)acrylic acid, 98%, Thermo Scientific Chemicals
Catalog numberB22443.14
Price (USD)/ Each
696.00
-
Add to cart
Chemical Identifiers
CAS381-98-6
IUPAC Name2-(trifluoromethyl)prop-2-enoate
Molecular FormulaC4H2F3O2
InChI KeyVLSRKCIBHNJFHA-UHFFFAOYSA-M
SMILES[O-]C(=O)C(=C)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused solid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Melting Point (clear melt)47.0-54.0?C
2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Jun Matsui.; Otto Doblhoff-Dier.; Toshifumi Takeuchi. 2-(Trifluoromethyl)acrylic acid: a novel functional monomer in non-covalent molecular imprinting. Analytica Chimica Acta. 1997, 343, (1-2), 1-4.
  2. Jun Matsui.; Hiroyuki Kubo.; Toshifumi Takeuchi. Molecularly Imprinted Fluorescent-Shift Receptors Prepared with 2-(Trifluoromethyl)acrylic Acid. Anal. Chem.2000, 72, (14), 3286-3290.