Thermo Scientific Chemicals

Salicylsalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22444.14
25 g, Each
Thermo Scientific Chemicals

Salicylsalicylic acid, 98%, Thermo Scientific Chemicals

Catalog number: B22444.14
25 g, Each
Quantity
Catalog number: B22444.14
also known as B22444-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
552-94-3
IUPAC Name
2-(2-hydroxybenzoyloxy)benzoic acid
Molecular Formula
C14H10O5
InChI Key
WVYADZUPLLSGPU-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=C1OC(=O)C1=CC=CC=C1O
Appearance (Color)
White
Form
Powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (Silylated GC)
≥97.5%
Identification (FTIR)
Conforms

Description

Salicylsalicylic acid is used in organic synthesis and functions as a plant hormone, as a key ingredient in topical anti-acne products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Salicylsalicylic acid is used in organic synthesis and functions as a plant hormone, as a key ingredient in topical anti-acne products.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text