Thermo Scientific Chemicals

2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals

Catalog number: B22445.06
5 g, Each
Thermo Scientific Chemicals

2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals

Catalog number: B22445.06
5 g, Each
Quantity
Catalog number: B22445.06
also known as B22445-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2035-72-5
IUPAC Name
4-benzoyl-3-hydroxyphenyl 2-methylprop-2-enoate
Molecular Formula
C17H14O4
InChI Key
IMNBHNRXUAJVQE-UHFFFAOYSA-N
SMILES
CC(=C)C(=O)OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
Appearance (Color)
Pale yellow
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%
Identification (FTIR)
Conforms
Solution Test
10% in ether: clear solution

Description

2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text