2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals
2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B22445.22
also known as B22445-22
Price (USD)/ Each
974.65
Online exclusive
1083.00 
Save 108.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
974.65
Online exclusive
1083.00 
Save 108.35 (10%)
Add to cart
2-Hydroxy-4-(methacryloyloxy)benzophenone, 99%, Thermo Scientific Chemicals
Catalog numberB22445.22
Price (USD)/ Each
974.65
Online exclusive
1083.00 
Save 108.35 (10%)
-
Add to cart
Chemical Identifiers
CAS2035-72-5
IUPAC Name4-benzoyl-3-hydroxyphenyl 2-methylprop-2-enoate
Molecular FormulaC17H14O4
InChI KeyIMNBHNRXUAJVQE-UHFFFAOYSA-N
SMILESCC(=C)C(=O)OC1=CC=C(C(=O)C2=CC=CC=C2)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.5%
Identification (FTIR)Conforms
Solution Test10% in ether: clear solution
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-(methacryloyloxy)benzophenone is used in ophthalmic devices, films, coatings, and fibers, cosmetic preparations and toners.

Solubility
Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
S. Nanjundan.; C.S.Jone Selvamalar.; R. Jayakumar. Synthesis and characterization of poly(3-acetyl-4-hydroxyphenyl acrylate) and its Cu(II) and Ni(II) complexes. European Polymer Journal. 2004, 40, (10), 2313-2321.