Thermo Scientific Chemicals

2-Fluoro-4-iodoaniline, 99%, Thermo Scientific Chemicals

Catalog number: B22450.22
100 g, Each
Thermo Scientific Chemicals

2-Fluoro-4-iodoaniline, 99%, Thermo Scientific Chemicals

Catalog number: B22450.22
100 g, Each
Quantity
Catalog number: B22450.22
also known as B22450-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
29632-74-4
IUPAC Name
2-fluoro-4-iodoaniline
Molecular Formula
C6H5FIN
InChI Key
CUMTUBVTKOYYOU-UHFFFAOYSA-N
SMILES
NC1=CC=C(I)C=C1F
Appearance (Color)
Pale cream to cream to pale brown to gray
Form
Powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
50.0-56.0?C

Description

2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-4-iodoaniline is widely used in the pharmaceutical, chemical and food industries as a fundamental building block to create a variety of important compounds.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text