Thermo Scientific Chemicals

4-(Methylthio)benzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: B22455.06
5 g, Each
Thermo Scientific Chemicals

4-(Methylthio)benzyl alcohol, 98%, Thermo Scientific Chemicals

Catalog number: B22455.06
5 g, Each
Quantity
Catalog number: B22455.06
also known as B22455-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3446-90-0
IUPAC Name
[4-(methylsulfanyl)phenyl]methanol
Molecular Formula
C8H10OS
InChI Key
MTXQKSQYMREAGJ-UHFFFAOYSA-N
SMILES
CSC1=CC=C(CO)C=C1
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Melting Point (clear melt)
38.5-44.5?C

Description

4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text