Thermo Scientific Chemicals

Tris(trimethylsilyl)silane, 97%, Thermo Scientific Chemicals

Catalog number: B22457.06
5 g, Each
Thermo Scientific Chemicals

Tris(trimethylsilyl)silane, 97%, Thermo Scientific Chemicals

Catalog number: B22457.06
5 g, Each
Quantity
Catalog number: B22457.06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1873-77-4
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4860-1.4925 @ 20?C

Description

Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(trimethylsilyl)silane is used in hydrosilylations of carbonyls, radical reactions, reductions of acid chlorides, and carbon-halogen bonds. It acts a mediator of radical reactions and considered to be a nontoxic substitute for tri-n-butylstannane in radical reactions. It forms complexes with transition metals and main group elements. Further, it is utilized in carbon-carbon bond forming reactions.

Solubility
Miscible with pentane, ether, toluene and tetrahydrofuran. Sparingly miscible with acetone and acetonitrile. Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text