Thermo Scientific Chemicals

1-Hexadecylamine, tech. 90%, remainder mainly 1-octadecylamine, Thermo Scientific Chemicals

Catalog number: B22459.30
250 g, Each
Thermo Scientific Chemicals

1-Hexadecylamine, tech. 90%, remainder mainly 1-octadecylamine, Thermo Scientific Chemicals

Catalog number: B22459.30
250 g, Each
Quantity
Catalog number: B22459.30
also known as B22459-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-27-1
IUPAC Name
hexadecan-1-amine
Molecular Formula
C16H35N
InChI Key
FJLUATLTXUNBOT-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCN
Appearance (Color)
White to cream or pale yellow
Form
Crystals or powder or crystalline powder or fused/lumpy solid or chunks
Assay (GC)
≥88.0%
Melting Point (clear melt)
38.0-49.0?C

Description

1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexadecylamine is used for producing resin, senior detergent, anti-caking agent.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents, acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text