o-Thiocresol is used as a thiol for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
o-Thiocresol is used as a thiol for proteomics research.
Solubility
Soluble in ether and ethanol. Insoluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only
E. Campaigne.; S. Osborn. Notes - Improved Procedure for the Preparation of Aromatic Thiols. J. Org. Chem. 1957, 22, (5), 561-562.
Yasser M. Riyad.; Ralf Hermann.; Ortwin Brede. GarnierHidden triplet states of aromatic thiols characterized by radiation-chemical sensitizations. Chemical Physics Letters. 2003, 379, (5-6), 776-780.