Ethyl thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals
Ethyl thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number B22468.14
also known as B22468-14
Price (USD)/ Each
454.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
454.00
Add to cart
Ethyl thiophene-3-carboxylate, 98%, Thermo Scientific Chemicals
Catalog numberB22468.14
Price (USD)/ Each
454.00
-
Add to cart
Chemical Identifiers
CAS5751-80-4
IUPAC Nameethyl thiophene-3-carboxylate
Molecular FormulaC7H8O2S
InChI KeyOYSLMAQEMAJMCL-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5205-1.5245 @ 20?C
FormLiquid
Assay (GC)≥97.5%
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-3-carboxylate is used as an intermediate in organic syntheses

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. A. D. Britt.; William B. Moniz. Reactivity of first-singlet excited xanthene laser dyes in solution. J. Org. Chem. 1973, 38, (5), 1057-1059.
  2. F. Demanze.; A. Yassar.; F. Garnier. Alternating Donor-Acceptor Substitutions in Conjugated Polythiophenes. Macromolecules. 1996, 29, (12), 4267-4273.