Thermo Scientific Chemicals

Imidazole-4-acetic acid monohydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B22473.03
1 g, Each
Thermo Scientific Chemicals

Imidazole-4-acetic acid monohydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: B22473.03
1 g, Each
Quantity
Catalog number: B22473.03
also known as B22473-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3251-69-2
IUPAC Name
2-(1H-imidazol-5-yl)acetic acid hydrochloride
Molecular Formula
C5H7ClN2O2
InChI Key
MWHLCFYPFGFBQO-UHFFFAOYSA-N
SMILES
Cl.OC(=O)CC1=CN=CN1
Appearance (Color)
Pale cream to cream to pale brown
Form
Crystals or powder or crystalline powder
Assay (Titration ex Chloride)
≥96.0 to ≤104.0%
Assay (HPLC)
≥96.0%

Description

A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A GABAc antagonistImidazole-4-acetic acid monohydrochloride is used in the preparation of imidazolyl-polyethylenimine modified nanoparticles. It is also used to prepare pyridyl and imidazoyl functionalized carboproteins and potential metal ion chelators. Further, it acts as an imidazole derivative with potential antituberculousis properties. In addition to this, it is used in the generation of acyl guanidine inhibitors of beta-secretas.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text