4-Hydroxy-3,5-dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
4-Hydroxy-3,5-dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-3,5-dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number B22474.03
also known as B22474-03
Price (USD)/ Each
65.70
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
65.70
Add to cart
4-Hydroxy-3,5-dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals
Catalog numberB22474.03
Price (USD)/ Each
65.70
-
Add to cart
Chemical Identifiers
CAS1019-52-9
IUPAC Name4-hydroxy-3,5-dinitrobenzoic acid
Molecular FormulaC7H4N2O7
InChI KeyGBSWIDSKAJFWMF-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)245.0-251.0?C
Appearance (Color)Yellow to pale brown
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only