Thermo Scientific Chemicals

4-Hydroxy-3,5-dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B22474.06
5 g, Each
Thermo Scientific Chemicals

4-Hydroxy-3,5-dinitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B22474.06
5 g, Each
Quantity
Catalog number: B22474.06
also known as B22474-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1019-52-9
IUPAC Name
4-hydroxy-3,5-dinitrobenzoic acid
Molecular Formula
C7H4N2O7
InChI Key
GBSWIDSKAJFWMF-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O
Melting Point (clear melt)
245.0-251.0?C
Form
Powder
Appearance (Color)
Yellow to pale brown
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text