Thermo Scientific Chemicals

3-Aminosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22478.02
0.5 g, Each
Thermo Scientific Chemicals

3-Aminosalicylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B22478.02
0.5 g, Each
Quantity
Catalog number: B22478.02
also known as B22478-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
570-23-0
IUPAC Name
3-amino-2-hydroxybenzoic acid
Molecular Formula
C7H7NO3
InChI Key
IQGMRVWUTCYCST-UHFFFAOYSA-N
SMILES
NC1=CC=CC(C(O)=O)=C1O
Comment
Purchased in U.K. and U.S.
Identification (FTIR)
Conforms (non-U.S. specification)
Assay (Silylated GC)
≥96.0%
Melting Point
232-242°C (U.S. specification)
Appearance (Color)
White to cream or green to brown to grey

Description

3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.

Solubility
Soluble in potassium hydroxide. Slightly soluble in water.

Notes
Air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text