3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Aminosalicylic acid acts as an oxygen, nitrogen donor ligand and forms complexes with silver.
Solubility
Soluble in potassium hydroxide. Slightly soluble in water.
Notes
Air and light sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Harscoat-Schiavo, C.; Neurohr, C.; Lecomte, S.; Marchivie, M.; Paternault, P. S. Influence of isomerism on recrystallization and cocrystallization induced by CO2 as an antisolvent. CrystEngComm 2015, 17 (29), 5410-5421.
Singh, A. K.; Singh, M. QCM sensing of melphalan via electropolymerized molecularly imprinted polythiophene films. Biosens. Bioelectron. 2015, 74, 711-717.