Thermo Scientific Chemicals

Phosphorus(V) bromide, 95%, Thermo Scientific Chemicals

Catalog number: B22481.22
100 g, Each
Thermo Scientific Chemicals

Phosphorus(V) bromide, 95%, Thermo Scientific Chemicals

Catalog number: B22481.22
100 g, Each
Quantity
Catalog number: B22481.22
also known as B22481-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-69-7
IUPAC Name
pentabromo-λ⁵-phosphane
Molecular Formula
Br5P
InChI Key
QRKVRHZNLKTPGF-UHFFFAOYSA-N
SMILES
BrP(Br)(Br)(Br)Br
Appearance (Color)
Yellow to orange
Form
Crystalline powder and/or Lumps
Assay (Titration ex Bromide)
>94.0 to <106.0%

Description

Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphorus(V) bromide, is used in organic chemistry to convert carboxylic acids to acyl bromides

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text