2-Chloro-2',4'-difluoroacetophenone, 98%, Thermo Scientific Chemicals
2-Chloro-2',4'-difluoroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloro-2',4'-difluoroacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
25 g
Catalog number B22487.14
also known as B22487-14
Price (USD)/ Each
199.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
199.00
Add to cart
2-Chloro-2',4'-difluoroacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberB22487.14
Price (USD)/ Each
199.00
-
Add to cart
Chemical Identifiers
CAS51336-94-8
IUPAC Name2-chloro-1-(2,4-difluorophenyl)ethan-1-one
Molecular FormulaC8H5ClF2O
InChI KeyUENGBOCGGKLVJJ-UHFFFAOYSA-N
SMILESFC1=CC=C(C(=O)CCl)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)45.0-52.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only