Thermo Scientific Chemicals

Methyl 2,5-dichlorobenzoate, 99%, Thermo Scientific Chemicals

Catalog number: B22489.09
10 g, Each
Thermo Scientific Chemicals

Methyl 2,5-dichlorobenzoate, 99%, Thermo Scientific Chemicals

Catalog number: B22489.09
10 g, Each
Quantity
Catalog number: B22489.09
also known as B22489-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2905-69-3
IUPAC Name
methyl 2,5-dichlorobenzoate
Molecular Formula
C8H6Cl2O2
InChI Key
SPJQBGGHUDNAIC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(Cl)=CC=C1Cl
Appearance (Color)
White to cream
Form
Crystalline powder or lumpy powder or fused solid
Assay (GC)
≥98.5%
Melting Point (clear melt)
37-42?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text