Thermo Scientific Chemicals

Magnesium silicate, activated, for chromatography, 60-100 mesh, Thermo Scientific Chemicals

Catalog number: B22490.30
250 g, Each
Thermo Scientific Chemicals

Magnesium silicate, activated, for chromatography, 60-100 mesh, Thermo Scientific Chemicals

Catalog number: B22490.30
250 g, Each
Quantity
Catalog number: B22490.30
also known as B22490-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1343-88-0
IUPAC Name
silicon(4+) magnesium(2+) trioxidandiide
Molecular Formula
MgO3Si
InChI Key
ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol)
100.39
Appearance (Color)
White to pale cream
Form
Powder
Particle Size
60-100 mesh (0.140-0.250mm): min. 90%

Description

Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium silicate, activated, is used in polyether adsorption and decolorization effect. Also used in chromatography.

Solubility
Insoluble in water.

Notes
Incompatible with strong acids and hydrogen fluoride.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text