Thermo Scientific Chemicals

2-Bromopropionyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: B22504.36
500 g, Each
Thermo Scientific Chemicals

2-Bromopropionyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: B22504.36
500 g, Each
Quantity
Catalog number: B22504.36
also known as B22504-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
563-76-8
IUPAC Name
2-bromopropanoyl bromide
Molecular Formula
C3H4Br2O
InChI Key
ILLHORFDXDLILE-UHFFFAOYNA-N
SMILES
CC(Br)C(Br)=O
Appearance (Color)
Clear colorless to yellow or pink
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.5175-1.5225 @ 20?C

Description

2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromopropionyl bromide is used in the preparation of cellulose macroinitiator through direct acylation of cellulose in ionic liquid 1-allyl-3-methylimidazolium chloride. It is also used to prepare 3-(2-bromopropionyl)-2-oxazolidone derivatives and bromoester terminated poly(3-hexylthiophene). Further, it is used as a macroinitiator for atom transfer radical polymerization of acrylates.

Solubility
Miscible with trichloromethane, diethyl ether, benzene and acetone.

Notes
Incompatible with water, alcohols, oxidizing agents and strong alkalis.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text