Thermo Scientific Chemicals

5-Hexen-2-one, 98%, Thermo Scientific Chemicals

Catalog number: B22505.14
25 g, Each
Thermo Scientific Chemicals

5-Hexen-2-one, 98%, Thermo Scientific Chemicals

Catalog number: B22505.14
25 g, Each
Quantity
Catalog number: B22505.14
also known as B22505-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-49-9
IUPAC Name
hex-5-en-2-one
Molecular Formula
C6H10O
InChI Key
RNDVGJZUHCKENF-UHFFFAOYSA-N
SMILES
CC(=O)CCC=C
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4190-1.4220 @ 20?C
Appearance (Color)
Clear colorless to pale yellow

Description

Allyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl Acetone is used as a nature identical flavor and fragrance. And it is also used to produce 2-methyl-hex-5-en-2-ol.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text