Thermo Scientific Chemicals

(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%, Thermo Scientific Chemicals

Catalog number: B22506.06
5 g, Each
Thermo Scientific Chemicals

(+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%, Thermo Scientific Chemicals

Catalog number: B22506.06
5 g, Each
Quantity
Catalog number: B22506.06
also known as B22506-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88568-95-0
IUPAC Name
methyl (2S)-2-{[(benzyloxy)carbonyl]amino}-2-(dimethoxyphosphoryl)acetate
Molecular Formula
C13H18NO7P
InChI Key
GSYSFVSGPABNNL-NSHDSACASA-N
SMILES
COC(=O)[C@@H](NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
Assay (unspecified)
(by Carbon EA) : >96.0 to<104.0%
Elemental Analysis
(Carbon) : 45.25-49.01% (Theory 47.14%)
Elemental Analysis
(Hydrogen) : 5.25-5.69% (Theory 5.48%)
Proton NMR
Conforms to structure
Phosphorus-31 NMR
Conforms to structure

Description

(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text