Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals

Catalog number: B22515.14
25 g, Each
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)-2,2',2″-nitrilotriethanol, 98%, Thermo Scientific Chemicals

Catalog number: B22515.14
25 g, Each
Quantity
Catalog number: B22515.14
also known as B22515-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6976-37-0
IUPAC Name
2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C8H19NO5
InChI Key
OWMVSZAMULFTJU-UHFFFAOYSA-N
SMILES
OCCN(CCO)C(CO)(CO)CO
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5% to ≤102.5%
Identification (FTIR)
Conforms
Appearance (Color)
White to pale cream

Description

It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a biological buffer, organic synthesize, chemical & pharmaceutical intermediate. It is suitable for fractionation of hemoglobin.

Solubility
Soluble in water (209.2 g/L), at 20°C and alcohol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text