Thermo Scientific Chemicals

(3-Chloropropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: B22517.22
100 g, Each
Thermo Scientific Chemicals

(3-Chloropropyl)trimethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: B22517.22
100 g, Each
Quantity
Catalog number: B22517.22
also known as B22517-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2530-87-2
IUPAC Name
(3-chloropropyl)trimethoxysilane
Molecular Formula
C6H15ClO3Si
InChI Key
OXYZDRAJMHGSMW-UHFFFAOYSA-N
SMILES
CO[Si](CCCCl)(OC)OC
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Refractive Index
1.4180-1.4230 @ 20?C

Description

(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Chloropropyl)trimethoxysilane is a good coupling agent for epoxy resins to glass in laminates.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, water/ moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text