Methyl 4-amino-3-chlorobenzoate, 99%, Thermo Scientific Chemicals
Methyl 4-amino-3-chlorobenzoate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-amino-3-chlorobenzoate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B22519.06
also known as B22519-06
Price (USD)/ Each
165.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
165.00
Add to cart
Methyl 4-amino-3-chlorobenzoate, 99%, Thermo Scientific Chemicals
Catalog numberB22519.06
Price (USD)/ Each
165.00
-
Add to cart
Chemical Identifiers
CAS84228-44-4
IUPAC Namemethyl 4-amino-3-chlorobenzoate
Molecular FormulaC8H8ClNO2
InChI KeyWKVJBYKFGNVWLM-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(N)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Melting Point (clear melt)105.0-111.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only