Thermo Scientific Chemicals

3-Amino-9-ethylcarbazole, 95%, Thermo Scientific Chemicals

Catalog number: B22529.22
100 g, Each
Thermo Scientific Chemicals

3-Amino-9-ethylcarbazole, 95%, Thermo Scientific Chemicals

Catalog number: B22529.22
100 g, Each
Quantity
Catalog number: B22529.22
also known as B22529-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
132-32-1
IUPAC Name
9-ethyl-9H-carbazol-3-amine
Molecular Formula
C14H14N2
InChI Key
OXEUETBFKVCRNP-UHFFFAOYSA-N
SMILES
CCN1C2=C(C=CC=C2)C2=C1C=CC(N)=C2
Comment
Specification differs for U.S. and non-U.S. material where indicated
Form
Powder or lumps
Appearance (Color)
Yellow to brown
Assay (GC)
≥94.0% (non-U.S. specification)
Assay from Suppliers CofA
≥94.0% (U.S. specification)

Description

Peroxidase substrate for immunoblotting and immunohistochemical staining procedures

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text