4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Benzoyl-3-methyl-1-phenyl-5-pyrazolinone, 98+%, Thermo Scientific Chemicals

Have Questions?
Quantity:
50 g
Catalog number B22530.18
also known as B22530-18
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS4551-69-3
IUPAC Name4-benzoyl-3-methyl-1-phenyl-4,5-dihydro-1H-pyrazol-5-one
Molecular FormulaC17H14N2O2
InChI KeyJDOLPAMOKSAEMD-UHFFFAOYNA-N
SMILESCC1=NN(C(=O)C1C(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Melting Point (clear melt)88.0-95.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only