Thermo Scientific Chemicals

Methyl 4-hydroxyphenylacetate, 98+%, Thermo Scientific Chemicals

Catalog number: B22533.14
25 g, Each
Thermo Scientific Chemicals

Methyl 4-hydroxyphenylacetate, 98+%, Thermo Scientific Chemicals

Catalog number: B22533.14
25 g, Each
Quantity
Catalog number: B22533.14
also known as B22533-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14199-15-6
IUPAC Name
methyl 2-(4-hydroxyphenyl)acetate
Molecular Formula
C9H10O3
InChI Key
XGDZEDRBLVIUMX-UHFFFAOYSA-N
SMILES
COC(=O)CC1=CC=C(O)C=C1
Melting Point (clear melt)
54-59?C
Appearance (Color)
White to cream or pale yellow
Form
Crystals or crystalline powder
Assay (Silylated GC)
≥98.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text