Strontium bromide hexahydrate, 99%, Thermo Scientific Chemicals
Strontium bromide hexahydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Strontium bromide hexahydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number B22553.22
also known as B22553-22
Price (USD)/ Each
60.65
Online exclusive
67.70 
Save 7.05 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
60.65
Online exclusive
67.70 
Save 7.05 (10%)
Add to cart
Strontium bromide hexahydrate, 99%, Thermo Scientific Chemicals
Catalog numberB22553.22
Price (USD)/ Each
60.65
Online exclusive
67.70 
Save 7.05 (10%)
-
Add to cart
Chemical Identifiers
CAS7789-53-9
IUPAC Namestrontium(2+) hexahydrate dibromide
Molecular FormulaBr2H12O6Sr
InChI KeyFLMJUJXBFKFYOZ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Br-].[Br-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Bromide)≥98.5 to ≤101.5%
Strontium bromide hexahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Strontium bromide hexahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water (2850 g/L at 20°C).

Notes
m.p. 88° (rapidly heated) , 180° -6H{2}O
RUO – Research Use Only

General References:

  1. E.Buzagh-Gere; J.Simon; S.Gal. Characterization of differently bound water by means of the constants of thermal decomposition kinetics. Fresenius' Zeitschrift für analytische Chemie. 1973, 264,(5), 392-396.
  2. W.D Shuster; S.Subler; E.L.McCoy. The influence of earthworm community structure on the distribution and movement of solutes in a chisel-tilled soil. Applied Soil Ecology. 2002, 21,(2), 159-167.