N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester, 97%, Thermo Scientific Chemicals
N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number B22557.06
also known as B22557-06
Price (USD)/ Each
91.50
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
91.50
Add to cart
N-Benzyloxycarbonyl-L-glutamic acid 5-tert-butyl ester, 97%, Thermo Scientific Chemicals
Catalog numberB22557.06
Price (USD)/ Each
91.50
-
Add to cart
Chemical Identifiers
CAS3886-08-6
IUPAC Name(2S)-2-{[(benzyloxy)carbonyl]amino}-5-(tert-butoxy)-5-oxopentanoate
Molecular FormulaC17H22NO6
InChI KeyGLMODRZPPBZPPB-ZDUSSCGKSA-M
SMILESCC(C)(C)OC(=O)CC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)84.0-96.0?C
Optical Rotation-13.3 ± 0.1? (C=2 in methanol)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only