Thermo Scientific Chemicals

Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B22559.14
25 g, Each
Thermo Scientific Chemicals

Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals

Catalog number: B22559.14
25 g, Each
Quantity
Catalog number: B22559.14
also known as B22559-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6779-09-5
IUPAC Name
ethylphosphonate
Molecular Formula
C2H5O3P
InChI Key
GATNOFPXSDHULC-UHFFFAOYSA-L
SMILES
CCP([O-])([O-])=O
Assay from Suppliers CofA
≥98.0%
Appearance (Color)
White to off-white
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
57.0-66.0°C

Description

Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text